missing translation for 'onlineSavingsMsg'
Learn More

Pyrilamine Maleate 98.0+%, TCI America™

Catalog No. p-7135544
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23695G 5 g
P236925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23695G Supplier TCI America Supplier No. P23695G
Only null left

Chemical Identifiers

CAS 59-33-6
Molecular Formula C21H27N3O5
Molecular Weight (g/mol) 401.46
MDL Number MFCD00069333
InChI Key JXYWFNAQESKDNC-BTJKTKAUSA-N
Synonym pyrilamine maleate, mepyramine maleate, anisopyradamine, histosol, minihist, diaminide maleate, paraminyl maleate, antihist, histatex, histine
PubChem CID 5284451
IUPAC Name (2Z)-but-2-enedioic acid; N-[2-(dimethylamino)ethyl]-N-[(4-methoxyphenyl)methyl]pyridin-2-amine
SMILES OC(=O)\C=C/C(O)=O.COC1=CC=C(CN(CCN(C)C)C2=CC=CC=N2)C=C1

Specifications

Melting Point 102°C
Color White
Quantity 5 g
Formula Weight 401.46
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyrilamine Maleate
RTECSNumber UT1225000
TSCA Yes