Learn More
Pyridoxamine dihydrochloride, Cell Culture Reagent
CAS: 524-36-7 | C8H14Cl2N2O2 | 241.112 g/mol
Supplier: Thermo Scientific Chemicals J6267906
Description
A vitamin in the Vitamin B-6 family
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Pyridoxamine dihydrochloride | |
| 220°C to 224°C | |
| C8H14Cl2N2O2 | |
| 5 g | |
| pyridoxamine dihydrochloride, 4-aminomethyl-5-hydroxymethyl-2-methylpyridin-3-ol dihydrochloride, pyridoxamine dichlorohydrate, pyridoxylamine dihydrochloride, 4-aminomethyl-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride, unii-yq8ncr7v0o, pyridoxamine 2hcl, yq8ncr7v0o, 2-methyl-3-hydroxy-4-aminomethyl-5-hydroxymethylpyridene dihydrochloride, pyridoxemine, dihydrochloride | |
| HNWCOANXZNKMLR-UHFFFAOYSA-N | |
| 4-(aminomethyl)-5-(hydroxymethyl)-2-methylpyridin-3-ol;dihydrochloride | |
| 10664 | |
| Solid |
| 524-36-7 | |
| Glass bottle | |
| MFCD00012808 | |
| 3632748 | |
| Soluble in DMSO,water,or methanol /n | |
| CC1=NC=C(C(=C1O)CN)CO.Cl.Cl | |
| 241.112 | |
| 241.12 |
Chemical Identifiers
| 524-36-7 | |
| 241.112 | |
| HNWCOANXZNKMLR-UHFFFAOYSA-N | |
| 10664 | |
| CC1=NC=C(C(=C1O)CN)CO.Cl.Cl |
| C8H14Cl2N2O2 | |
| MFCD00012808 | |
| pyridoxamine dihydrochloride, 4-aminomethyl-5-hydroxymethyl-2-methylpyridin-3-ol dihydrochloride, pyridoxamine dichlorohydrate, pyridoxylamine dihydrochloride, 4-aminomethyl-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride, unii-yq8ncr7v0o, pyridoxamine 2hcl, yq8ncr7v0o, 2-methyl-3-hydroxy-4-aminomethyl-5-hydroxymethylpyridene dihydrochloride, pyridoxemine, dihydrochloride | |
| 4-(aminomethyl)-5-(hydroxymethyl)-2-methylpyridin-3-ol;dihydrochloride |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-357-6
RTECSNumber : UV1230000
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only