missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxamine dihydrochloride, Cell Culture Reagent

Catalog No. p-7076593
Change view
Click to view available options
Quantity:
2.5 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6267927 2.5 g
AAJ6267906 5 g
AAJ6267909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6267927 Supplier Thermo Scientific Chemicals Supplier No. J6267927

CAS: 524-36-7 | C8H14Cl2N2O2 | 241.112 g/mol

A vitamin in the Vitamin B-6 family

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 524-36-7
Molecular Formula C8H14Cl2N2O2
Molecular Weight (g/mol) 241.112
MDL Number MFCD00012808
InChI Key HNWCOANXZNKMLR-UHFFFAOYSA-N
Synonym pyridoxamine dihydrochloride, 4-aminomethyl-5-hydroxymethyl-2-methylpyridin-3-ol dihydrochloride, pyridoxamine dichlorohydrate, pyridoxylamine dihydrochloride, 4-aminomethyl-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride, unii-yq8ncr7v0o, pyridoxamine 2hcl, yq8ncr7v0o, 2-methyl-3-hydroxy-4-aminomethyl-5-hydroxymethylpyridene dihydrochloride, pyridoxemine, dihydrochloride
PubChem CID 10664
IUPAC Name 4-(aminomethyl)-5-(hydroxymethyl)-2-methylpyridin-3-ol;dihydrochloride
SMILES CC1=NC=C(C(=C1O)CN)CO.Cl.Cl

Specifications

Melting Point 220°C to 224°C
Packaging Glass bottle
Quantity 2.5 g
Beilstein 3632748
Solubility Information Soluble in DMSO,water,or methanol /n
Formula Weight 241.12
Physical Form Solid
Chemical Name or Material Pyridoxamine dihydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-357-6
RTECSNumber UV1230000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only