Learn More
Corning™ PYREX™ Shorty Disposable Glass Serological Pipets, TD, Sterile, Plugged
Ideal for use in confined areas such as hoods
$809.38 - $3313.28
Specifications
Aspirator Plug Yes
Calibrated Yes
Material Borosilicate Glass
Graduated Yes
Tip Style Round
View More Specs
Products 8
Catalog Number Mfr. No. Volume (Metric) Graduations Negative Graduations Packaging Type Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Graduations Negative Graduations Packaging Type Price Quantity  
13-664-18D
Accessories View Documents
Corning™
7078B25
25 mL 0.2 mL 5 mL Multi-pack
Case of 400 Each for $3,313.28
Only null left
13-664-18C
Accessories View Documents
Corning™
7078B10
10 mL 0.2 mL 4 mL Multi-pack
Case of 400 Each for $2,305.94
Only null left
13-664-18A
Accessories View Documents
Corning™
7078B1
1 mL 0.01 mL 0.5 mL Multi-pack
Case of 500 Each for $1,222.94
Only null left
13-676-29H
Accessories View Documents
Corning™
7077B10
10 mL 0.2 mL 4 mL Individually Wrapped
Case of 400 Each for $2,376.51
Only null left
13-676-29E
Accessories View Documents
Corning™
7077B50
50 mL 0.5 mL 3 mL Individually Wrapped
Case of 50 Each for $809.38
Only null left
13-676-29K
Accessories View Documents
Corning™
7077B1
1 mL 0.01 mL 0.5 mL Individually Wrapped
Case of 500 Each for $1,236.69
Only null left
13-676-29J
Accessories View Documents
Corning™
7077B5
5 mL 0.1 mL 3 mL Individually Wrapped
Case of 400 Each for $2,157.67
Only null left
13-676-29G
Accessories View Documents
Corning™
7077B25
25 mL 0.2 mL 5 mL Individually Wrapped
Case of 400 Each for $3,225.79
Only null left
Description
Description
These shorty pipets, calibrated 'To Deliver' with blow-out, are ideal for use in confined areas. The 5, 10, and 25 mL sizes have tooled tops. Negative graduations on each pipet make it applicable for both serological and measuring procedures. Packaged in plastic bags so several can be removed without contaminating the entire pack. ISO color-coding is printed directly on the tear strip.
Do not pipet by mouth. We suggest using a mechanical pipetting device.
Cat. No. 7078B-50 features negative and reverse graduations.
Compliance
Calibrated To Deliver (TD) with blow-out.
Specifications
Specifications
Yes
Borosilicate Glass
Round
Single-use
Yes
Yes
Serological Pipet
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'