Learn More
Corning™ PYREX™ Shorty Disposable Glass Serological Pipets, TD, Sterile, Plugged
Ideal for use in confined areas such as hoods
Supplier:  Corning™ 7077B25
 View more versions of this product
Catalog No. 13-676-29G
Only null left
Explore available promotions
Description
Description
These shorty pipets, calibrated 'To Deliver' with blow-out, are ideal for use in confined areas. The 5 mL, 10 mL, and 25 mL sizes have tooled tops. Negative graduations on each pipet make it applicable for both serological and measuring procedures. ISO color coding is printed directly on the Steri-ViewTM wrapper.
Do not pipet by mouth. We suggest using a mechanical pipetting device.
Cat. No. 7077B-50 features negative and reverse graduations.
Specifications
Specifications
Yes
To Deliver
Yes
11.8 in.
Round
Individually Wrapped
Red
Serological Pipet
Yes
Borosilicate Glass
0.2 mL
5 mL
Tooled
0.84 oz.
25 mL
Single-use
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories