Learn More
Corning™ PYREX™ HPLC Reservoir, Single Cavity
Reservoirs have a central cavity for a mobile phase inlet reservoir
Supplier: Corning™ 21505L
Description
Reservoirs have a central cavity for a mobile phase inlet reservoir
PYREX HPLC reservoirs have a central cavity for a mobile phase inlet reservoir, an autoclavable plastic coat that absorbs UV up to 385 nm, and a GL 45 thread finish. These may be used with our 3-hole cap (Cat. No. 1395-3H).
Order Info
Optional 3-hole cap available separately (13-645-099)
Specifications
| 7.6 in. | |
| Plastic | |
| Reservoirs have a central cavity for a mobile phase inlet reservoir | |
| 385 mm | |
| 1/Pk. | |
| 195 mm |
| 5 L | |
| HPLC Reservoir | |
| 15.15 in. | |
| 1/Cs. | |
| Autoclavable |