missing translation for 'onlineSavingsMsg'
Learn More

Pyranine

Catalog No. AAL1125206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1125206 5 g
AAL1125214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1125206 Supplier Thermo Scientific Chemicals Supplier No. L1125206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6358-69-6 | C16H7Na3O10S3 | 524.37 g/mol

Pyranine is used as a fluorescent pH indicator with emission change in the physiological pH range. It acts as a dye in optical sensor applications. It finds application as a coloring agent and biological stain. It is also used in the measurement of pH of intracellular fluid in the cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyranine is used as a fluorescent pH indicator with emission change in the physiological pH range. It acts as a dye in optical sensor applications. It finds application as a coloring agent and biological stain. It is also used in the measurement of pH of intracellular fluid in the cells.

Solubility
Soluble in water and glacial acetic acid.

Notes
This dye is a highly water soluble, brightly fluorescent dye useful as a fluorescent pH indicator with emission change in the physiological pH range and as a dye in optical sensor applications.Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 6358-69-6
Molecular Formula C16H7Na3O10S3
Molecular Weight (g/mol) 524.37
MDL Number MFCD00037575
InChI Key KXXXUIKPSVVSAW-UHFFFAOYSA-K
Synonym pyranine, solvent green 7, c.i. solvent green 7, japan green 204, hpts, FD&C green no. 8, japan green no. 204, pyrene 1, pyranine 120, FD&C green 8
PubChem CID 61388
ChEBI CHEBI:52083
IUPAC Name trisodium;8-hydroxypyrene-1,3,6-trisulfonate
SMILES [Na+].[Na+].[Na+].OC1=CC(=C2C=CC3=C(C=C(C4=CC=C1C2=C34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Quantity 5 g
Beilstein 4107272
Solubility Information Soluble in water and glacial acetic acid.
Formula Weight 524.39
Chemical Name or Material Pyranine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-783-6
RTECSNumber UR2700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only