missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Pyr3

Catalog No. 375150
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
375150 50 mg
375110 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 375150 Supplier Tocris Bioscience™ Supplier No. 3751/50*

Selective TRPC3 blocker

Selective antagonist of the canonical transient receptor potential channel 3 (TRPC3). Inhibits TRPC3-mediated Ca2+ influx (IC50 = 0.7 μM) and suppresses activation of nuclear factor of activated T cells (NFAT). Inhibits hypertrophic responses in cardiomyocytes.

Chemical Identifiers

CAS 1160514-60-2
Molecular Formula C16H11Cl3F3N3O3

Specifications

Quantity 50 mg
Formula Weight 456.63
Purity >98%
Molecular Weight (g/mol) 456.627
Chemical Name or Material Pyr3
Recommended Storage Store at -20°C