missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Pyr3
Description
Selective antagonist of the canonical transient receptor potential channel 3 (TRPC3). Inhibits TRPC3-mediated Ca2+ influx (IC50 = 0.7 μM) and suppresses activation of nuclear factor of activated T cells (NFAT). Inhibits hypertrophic responses in cardiomyocytes.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 456.63 |
| Purity | >98% |
| Molecular Weight (g/mol) | 456.627 |
| Chemical Name or Material | Pyr3 |
Safety and Handling
| Recommended Storage | Store at -20°C |