missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Purospher™ STAR RP-18 Endcapped (2 μm) Capillary Column, 0.3 mm I.D.
Description
- Intended to be used in the separation of aflatoxins, aldehydes, alkaloids, antibiotics, catecholamines, explosives, oligonucleotides, esters, fat soluble vitamins, lipids, fatty acids, flavonoids, ketones, nitrosamines, nucleosides, PAHs, PCBs, peptides, pesticides, phenols, phthalates, preservatives, steroids, metabolized steroids, sulfonamides, sweeteners, and water soluble vitamins.
Specifications
Specifications
| Diameter (Metric) | 0.3 mm |
| Length (Metric) | 50 mm |
| Particle Size | 2 μm |
| Pore Size | 200 Å |
| Stationary Phase | C18 (octadecyl) |
| Product Line | Purospher STAR |
| Phase | Reversed Phase |
| Material | Stainless Steel |
| Technique | UHPLC, LC/MS |
| Product Type | HPLC/UHPLC/UHPLC-MS Column |