missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Puritan™ UniTranz-RT Transport Systems
For the collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma
$1084.67 - $1609.12
Specifications
| Product Type | UniTranz-RT Transport System |
|---|---|
| Media Type | Hank's Balanced Salts, Bovine Serum Albumin, Gelatin, Sucrose, L-Cysteine, L-Glutamic Acid, Colistin, Amphotericin B, Vancomycin, Sodium Bicarbonate and HEPES Sodium Salt |
| Sterility | Sterile |
| Certifications/Compliance | CE |
| For Use With (Application) | Collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma |
| Catalog Number | Mfr. No. | Fill Volume | SwabConfiguration | Shaft Material | Tip Type | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Fill Volume | SwabConfiguration | Shaft Material | Tip Type | Price | Quantity | ||
|
22-027-100
|
Puritan™
UT106 |
1 mL | Single Applicator | Polystyrene | Large Flocked |
Case of 300 Each for $1,096.34
|
|
||
|
22-027-103
|
Puritan™
UT306 |
3 mL | Single Applicator | Polystyrene | Elongated Flocked |
Case of 300 Each for $1,496.54
|
|
||
|
22-027-102
|
Puritan™
UT117 |
1 mL | Single Applicator | Polystyrene | Ultrafine Flocked |
Case of 300 Each for $1,084.67
|
|
||
|
22-027-105
|
Puritan™
PURUT317 |
3 mL | Single Applicator | Polystyrene | Ultrafine Flocked |
Case of 300 Each for $1,415.74
|
|
||
|
22-027-108
|
Puritan™
UT302 |
3 mL | Dual Applicator | Aluminum, Plastic | 1 Standard, 1 Minitip |
Case of 300 Each for $1,489.54
|
|
||
|
22-027-106
|
Puritan™
UT367 |
3 mL | Dual Applicator | Polystyrene | Elongated, Ultrafine Flocked |
Case of 300 Each for $1,535.92
|
|
||
|
22-027-109
|
Puritan™
UT366 |
3 mL | Dual Applicator | Polystyrene | 1 Standard, 1 Minitip |
Case of 300 Each for $1,609.12
|
|
||
Description
For the collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma
Advanced kit design is easy to use and reliable.
- Ready-to-use
- Room temperature stable medium
- Leakproof self-centering cap, facilitates easy swab removal and disposal
- High-performance swab tip
- PurFlock™ Ultra swab with molded breakpoint designed to accommodate different sampling sites fast, safe and convenient
- Soft tip for patient comfort
- Premium medical grade plastic components to improve product shelf life
- Shelf life: 18 months
Specifications
| UniTranz-RT Transport System | |
| Sterile | |
| Collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma | |
| 7.3 ±0.2 | |
| 2°C to 25°C |
| Hank's Balanced Salts, Bovine Serum Albumin, Gelatin, Sucrose, L-Cysteine, L-Glutamic Acid, Colistin, Amphotericin B, Vancomycin, Sodium Bicarbonate and HEPES Sodium Salt | |
| CE | |
| Individually-wrapped | |
| 300 Cs. |