missing translation for 'onlineSavingsMsg'
Learn More

Puritan™ UniTranz-RT Transport Systems

Catalog No. 22027100
Change view
Click to view available options
Fill Volume:
1 mL
3 mL
Swab Configuration:
Dual Applicator
Single Applicator
Shaft Material:
Aluminum, Plastic
Polystyrene
Tip Type:
1 Standard, 1 Minitip
Elongated Flocked
Elongated, Ultrafine Flocked
Large Flocked
Ultrafine Flocked
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume Swab Configuration Shaft Material Tip Type
22027100 1 mL Single Applicator Polystyrene Large Flocked
22027103 3 mL Single Applicator Polystyrene Elongated Flocked
22027102 1 mL Single Applicator Polystyrene Ultrafine Flocked
22027105 3 mL Single Applicator Polystyrene Ultrafine Flocked
22027108 3 mL Dual Applicator Aluminum, Plastic 1 Standard, 1 Minitip
22027106 3 mL Dual Applicator Polystyrene Elongated, Ultrafine Flocked
22027109 3 mL Dual Applicator Polystyrene 1 Standard, 1 Minitip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 22027100 Supplier Puritan™ Supplier No. UT 106
Only null left

For the collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma

Intended for the collection and preservation of Viruses - Chlamydia - Mycoplasma and Ureaplasma; Contains antimicrobial agents to minimize bacterial and fungal contamination;Contains glass beads to release and disperse;Used in viral antigen detection test, PCR and rapid tests sample into the medium during vortexing; Used in viral antigen detection test, PCR and rapid tests; Available in a 1ml or 3ml fill, medium is stable at room temperature; Advanced kit design is easy to use and reliable; Self-centering cap is leakproof and facilitates easy swab removal and disposal; High-performance swab tip features patented PurFlock Ultra™ swab with molded breakpoint designed to accommodate different collection sites; Soft tip for patient comfort; Premium medical grade plastic components to improve product shelf life; All kits include sterile wrapped swabs and vial; Shelf life - 18 months; Made in USA

Specifications

Product Type UniTranz-RT Transport System
Media Type Hank's Balanced Salts, Bovine Serum Albumin, Gelatin, Sucrose, L-Cysteine, L-Glutamic Acid, Colistin, Amphotericin B, Vancomycin, Sodium Bicarbonate and HEPES Sodium Salt
Sterility Sterile
Swab Configuration Single Applicator
Certifications/Compliance CE
Fill Volume 1 mL
For Use With (Application) Collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma
Includes High performance large tip purFlock ultra swab, wrapped and 1mL Universal Transport Medium Filled Vial
Packaging Individually-wrapped
pH 7.3 ±0.2
Quantity 300 Cs.
Shaft Material Polystyrene
Storage Requirements 2°C to 25°C
Tip Type Large Flocked
Volume (Metric) 1 mL
Show More Show Less