missing translation for 'onlineSavingsMsg'
Learn More

Puritan™ UniTranz-RT Transport Systems

Catalog No. 22027105
Change view
Click to view available options
Fill Volume:
1 mL
3 mL
Swab Configuration:
Dual Applicator
Single Applicator
Shaft Material:
Aluminum, Plastic
Polystyrene
Tip Type:
1 Standard, 1 Minitip
Elongated Flocked
Elongated, Ultrafine Flocked
Large Flocked
Minitip Flocked
Ultrafine Flocked
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Fill Volume Swab Configuration Shaft Material Tip Type
22027105 3 mL Single Applicator Polystyrene Ultrafine Flocked
22027100 1 mL Single Applicator Polystyrene Large Flocked
22027101 1 mL Single Applicator Polystyrene Minitip Flocked
22027103 3 mL Single Applicator Polystyrene Elongated Flocked
22027102 1 mL Single Applicator Polystyrene Ultrafine Flocked
22027108 3 mL Dual Applicator Aluminum, Plastic 1 Standard, 1 Minitip
22027106 3 mL Dual Applicator Polystyrene Elongated, Ultrafine Flocked
22027109 3 mL Dual Applicator Polystyrene 1 Standard, 1 Minitip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 22027105 Supplier Puritan™ Supplier No. PURUT317

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For the collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma

Intended for the collection and preservation of Viruses - Chlamydia - Mycoplasma and Ureaplasma; Contains antimicrobial agents to minimize bacterial and fungal contamination;Contains glass beads to release and disperse;Used in viral antigen detection test, PCR and rapid tests sample into the medium during vortexing; Used in viral antigen detection test, PCR and rapid tests; Available in a 1ml or 3ml fill, medium is stable at room temperature; Advanced kit design is easy to use and reliable; Self-centering cap is leakproof and facilitates easy swab removal and disposal; High-performance swab tip features patented PurFlock Ultra™ swab with molded breakpoint designed to accommodate different collection sites; Soft tip for patient comfort; Premium medical grade plastic components to improve product shelf life; All kits include sterile wrapped swabs and vial; Shelf life - 18 months; Made in USA

Specifications

Product Type UniTranz-RT Transport System
Media Type Hank's Balanced Salts, Bovine Serum Albumin, Gelatin, Sucrose, L-Cysteine, L-Glutamic Acid, Colistin, Amphotericin B, Vancomycin, Sodium Bicarbonate and HEPES Sodium Salt
Sterility Sterile
Swab Configuration Single Applicator
Certifications/Compliance CE
Fill Volume 3 mL
For Use With (Application) Collection and preservation of Virus, Chlamydia, Mycoplasma and Ureaplasma
Includes High performance ultrafine purFlock ultra swab, wrapped and 3mL Universal Transport Medium Filled Vial
Packaging Individually-wrapped
pH 7.3 ±0.2
Quantity 300 Cs.
Shaft Material Polystyrene
Storage Requirements 2°C to 25°C
Tip Type Ultrafine Flocked
Show More Show Less