missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PureQuant B Cell Assay

Catalog No. A47196
Encompass
Change view
Click to view available options
Quantity:
28 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A47196 28 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A47196 Supplier Applied Biosystems™ Supplier No. A47196

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Applied Biosystems PureQuant assays are a family of assays that accurately identify and quantify specific immune cell types in a mixed population of cells. These assays are TaqMan qPCR-based and measure genomic DNA methylation at specific sites that serve as a unique identifier. The assays include standards and controls and each 96-well plate can accommodate up to seven test samples.

The Applied Biosystems PureQuant assays are a family of assays that accurately identify and quantify specific immune cell types in a mixed population of cells. These assays are TaqMan qPCR-based and measure genomic DNA methylation at specific sites that serve as a unique identifier. The assays include standards and controls and each 96-well plate can accommodate up to seven test samples. Results are provided as a percentage of target cell type out of total input cells using a free analysis tool.

The PureQuant B cell Assay utilizes the LRP5 gene as an epigenetic marker for B cells.

Benefits include:

  • Proven performance―core technology has been used for identification and quantification of immune cell types for over a decade
  • Easily standardized―qPCR method enables consistent results across multiple users, instruments, and sites
  • Facilitates regulatory filings―regulatory support file provided upon request; established performance specifications following ICHQ2(R1) guidelines
  • Scheduling flexibility―assay can be performed using fresh or frozen cells/gDNA

The assays include all reagents, standards, and controls required to run the assay. Specific components include:

  • Calibrator: corrects for variation in bisulfite conversion efficiency. Plasmid-based calibrator harbors the genomic target regions in equimolar ratios and includes GAPDH. Subjected to bisulfite conversion followed by copy number estimation via qPCR standard curve. Allows estimation of assay-specific bisulfite efficiency factor: the ratio between TpG/GAPDH copy numbers measured from the same plasmid.
  • Reference: checks assay performance. Consists of genomic DNA extracted from human blood of pooled healthy donors. The reference values represent the expected range of the target cell type. For example, CD8+ cells are generally more abundant than Treg or Th17 cells.
  • Standards: determine the unknown copy number. Consist of six dilutions spanning 4-log linear dynamic range (30-31250 copies). Absolute copy numbers can reliably be determined using the slope and intercept of the respective standard curves.

This product is manufactured at a site with an ISO9001-certified quality management system.

Specifications

Content And Storage • Dynabeads SILANE Genomic DNA Kit, store at 2°C to 8°C
• 1 x 2 mL PureQuant qPCR Master Mix, store at 2°C to 8°C
• 4 x ABS
• THFA, store at room temperature
• PureQuant B cell Module, store at -15° to -25°C (B cell and GAPDH TaqMan primers and probes, Reference DNA, Lambda DNA, Standard DNA)
Detection Method Primer-probe
Form Frozen
Format 96-Well Plate
PCR Method qPCR
Reaction Speed Fast or Standard
For Use With (Equipment) QuantStudio™
Product Type Purequant B Cell Assay
Quantity 28 reactions
No. of Reactions 28 reactions
Sample Type Cell
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.