missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ RNase A (20 mg/mL) DFS Item

PureLink™ RNase A (ribonuclease A) is a bovine pancreatic ribonuclease that cleaves single-stranded RNA.

$404.00 - $956.00

Specifications

Concentration 20 mg/mL
Packaging Type Bottle
Product Line UltraPure
Shipping Condition Room Temperature
Form Liquid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
12091021 DFS Item
View Documents
Invitrogen™
12091021
10 mL
Each for $404.00
Only null left
 
12091039 DFS Item
View Documents
Invitrogen™
12091039
25 mL
Each for $956.00
Only null left
 
Description

Description

  • RNase A (ribonuclease A) is a bovine pancreatic ribonuclease that cleaves single-stranded RNA
  • Used for the isolation of RNA-free DNA

Concentration

  • 20 mg RNase A/mL in 50 mM Tris-HCl (pH 8.0), 10 mM EDTA
Specifications

Specifications

20 mg/mL
UltraPure
Liquid
Bottle
Room Temperature
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.