missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ HiPure Expi Plasmid Megaprep Kit

Catalog No. K210008XP
Encompass
Change view
Click to view available options
Quantity:
4 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K210008XP 4 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K210008XP Supplier Invitrogen™ Supplier No. K210008XP
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The PureLink™ HiPure Plasmid Megaprep Kit also includes vacuum filtration columns that provide bacterial filtration without centrifugation, enabling extremely fast process times.

The PureLink™ HiPure Plasmid Megaprep Kit also includes vacuum filtration columns that provide bacterial filtration without centrifugation, enabling extremely fast process times. The PureLink™ HiPure Expi Plasmid Megaprep Kit typically isolates 4 mg of high quality, ultrapure plasmid DNA with inherently low endotoxin levels from 500 mL of bacterial culture.

Advantages of the PureLink™ HiPure Expi Plasmid Megaprep Purification Kit include:

  • High yield—isolate over 5mg of high quality plasmid DNA from a single purification using 1L of bacterial culture volume
  • Purity—inherently low endotoxin levels (typically 0.1–1.0 EU/μg) and A260/280 >1.8, making it ideal for mammalian cell transfection
  • Simple and super-fast protocol—with vacuum assisted protocol, typical plasmid isolation takes as little as 90 minutes


HiPure Expi anion exchange chromatography for plasmid purification
PureLink™ HiPure Expi plasmid purification kits use a novel, proprietary anion-exchange resin to enable the purification of the highest levels of transfection-grade plasmid DNA. The enhanced resin combines large capacity with fast flow rate, high resolution, and efficient endotoxin removal. Plasmid preparations are typically completed in less than 2 hours with purity equal to two CsCl gradients. PureLink™ HiPure Expi plasmid purification kits enable the quickest process for obtaining the highest amount of transfection-grade plasmid DNA.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Content And Storage • Equilibration Buffer
• Resuspension Buffer
• Lysis Buffer
• Precipitation Buffer
• RNase A
• Wash Buffer
• Elution Buffer
• TE Buffer
• Expi HiPure Mega Filtration Column
• Expi HiPure Mega DNA Binding Column

Store all components at room temperature upon receipt.
Format Kit
Isolation Technology Anion Exchange Resin
For Use With (Application) Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription
High-throughput Compatibility Manual Protocols
No. of Reactions 4 Preps
Prep Scale 4 mg plasmid DNA
Product Line PureLink
Product Type Plasmid MegaPrep Kit
Quantity 4 Preps
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Target Plasmid DNA
Test Time 2 hr.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.