missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Expi Endotoxin-Free Mega Plasmid Purification Kit

Catalog No. A31232
Encompass
Change view
Click to view available options
Quantity:
4 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31232 4 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A31232 Supplier Invitrogen™ Supplier No. A31232

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The PureLink™ Expi Endotoxin-Free Mega Plasmid Purification Kit enables quick and simple isolation of endotoxin-free plasmid DNA using an enhanced anion exchange resin.

The PureLink™ Expi Endotoxin-Free Mega Plasmid Purification Kit enables quick and simple isolation of endotoxin-free plasmid DNA using an enhanced anion exchange resin. Purify up to 5 mg of high quality plasmid DNA that is suitable for advanced transfection applications such as transfection of sensitive cells lines. The kit utilizes a novel vacuum-assisted purification workflow that dramatically reduces purification time to approximately 2 hours.

  • Simple and fast protocol—unlike other anion exchange-based endotoxin-free plasmid purification systems, a vacuum filtration method is used for lysate clarification and plasmid DNA purification. There is no waiting for gravity-flow columns, and purification typically takes 2 hours.
  • High yield—isolate up to 5 mg of high quality, endotoxin-free plasmid DNA
  • Endotoxin-free plasmid—produces endotoxin-free (less than 0.1 EU/μg) plasmid that is ideal for sensitive applications such as transfection of primary cells and research on gene therapy or plasmid vaccines
  • Purity—ultralow levels of protein, RNA, and genomic DNA contamination
  • Compatibility—purified plasmid is compatible with the Gibco™ ExpiCHO™ and Expi293™ transient mammalian expression systems
  • Endotoxin-free components—includes additional endotoxin-free elution buffer, water, and plasmid storage tubes

Specifications

Content And Storage

• Resuspension Buffer
• Lysis Indicator
• RNase A
• Lysis Buffer
• Precipitation Buffer
• Endotoxin Removal Buffer
• Wash Buffer
• Elution Buffer
• TE Buffer
• Expi Mega Filtration Column
• Expi Mega DNA Binding Column

Endotoxin-free components: elution buffer, water, plasmid storage tubes

Format Spin Column
Isolation Technology Anion Exchange Resin
Sample Type Bacterial Culture
Expression System Mammalian
For Use With (Application) Cloning, PCR
High-throughput Compatibility Manual Protocols
Prep Scale Large Volume Microbial Samples (> 10 mL)
Product Line PureLink
Product Type Plasmid Purification Kit
Quantity 4 preps
Shipping Condition Room Temperature
Target Endotoxin-free Plasmid
Test Time 2 hr.
Show More Show Less
What makes the PureLink Expi Endotoxin-Free Mega/Giga plasmid purification kits yield endotoxin-free plasmid DNA?

The anion exchange resin used in PureLink HiPure plasmid purification kits produce plasmid DNA with inherently low endotoxin levels of 0.1-1.0 EU/µg. To add to this, PureLink Expi Endotoxin-Free Mega/Giga plasmid purification kits contain endotoxin-free components, and a proprietary Endotoxin Removal Buffer that further enhances endotoxin depletion, resulting in plasmid DNA that is consistently endotoxin-free (< 0.1 EU/µg) .

Is the Endotoxin Removal Buffer (ER) in PureLink Expi Endotoxin-Free Mega/Giga plasmid purfiication kits sold separately?

No. Endotoxin Removal Buffer (ER) in PureLink Expi Endotoxin-Free Mega/Giga plasmid purification kits is not sold separately.

What is the endotoxin level in the plasmid DNA prepared using PureLink Expi Endotoxin-Free Mega/Giga plasmid purfiication kits?

The PureLink Expi Endotoxin-Free Mega/Giga plasmid purification kits can produce endotoxin-free (< 0.1 EU/µg) advanced transfection-grade plasmid DNA.

For research use only. Not for use in diagnostic procedures.