missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PureLink™ Expi Endotoxin-Free Buffer Set

Catalog No. A33074
Encompass
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A33074 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A33074 Supplier Invitrogen™ Supplier No. A33074
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Specifically, the buffer set includes RNAse A, Resuspension Buffer (R3), Lysis Buffer (L7), Precipitation Buffer (N3), Equilibration Buffer (EQ1), Wash Buffer (W8), Elution buffer (E4), TE Buffer (TE), Endotoxin Removal Buffer, Lysis Indicator, and endotoxin free water.

Specifically, the buffer set includes RNAse A, Resuspension Buffer (R3), Lysis Buffer (L7), Precipitation Buffer (N3), Equilibration Buffer (EQ1), Wash Buffer (W8), Elution buffer (E4), TE Buffer (TE), Endotoxin Removal Buffer, Lysis Indicator, and endotoxin free water.

  • Simple and super-fast protocol—Unlike other anion-exchange-based endotoxin free plasmid purification systems, the PureLink™ Expi Endotoxin-Free Maxi Plasmid Purification Kit utilizes vacuum filtration for lysate clarification and plasmid DNA purification. There is no waiting for gravity-flow columns and typical purification takes 90 minutes.
  • High yield—solate up to 1.5 mg of high quality, endotoxin-free plasmid DNA
  • Endotoxin-Free—Purify endotoxin-free (<0.1 EU/ μg) plasmid that is ideal for sensitive applications such as transfection of primary cells, genome engineering, and research on gene therapy or plasmid vaccines.
  • Purity—Ultralow levels of protein, RNA and genomic DNA contamination
  • Compatibility with ExpiCHO™ and Expi293™ Expression Systems—Plasmid purified using the PureLink™ Expi Endotoxin-Free Plasmid Purification Kit is compatible with the Gibco™ ExpiCHO™ and Expi293™ transient mammalian expression systems.

Specifications

Content And Storage • Resuspension buffer
• Lysis indicator
• RNase A
• Lysis buffer
• Precipitation buffer
• Endotoxin removal buffer
• Wash buffer
• Elution buffer
• TE buffer

Scale Maxi
Sample Type Bacteria
Expression System Mammalian
For Use With (Application) Cloning, PCR
High-throughput Compatibility Manual Protocols
Prep Scale > 1 mg (Large-Scale) Plasmid DNA, > 200 μg (Large-Scale) Plasmid DNA
Product Line PureLink
Product Type Buffer Set
Quantity 1 Set
Shipping Condition Room Temperature
Target Plasmid DNA
Test Time 90 min.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.