missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gentex™ PureFlo ESM+ PF60 Welding PAPR
Head mounted self-contained PAPR ideal for welding, fabrication, and radiant heat hazards.
Specifications
| ANSI Standard | Head Protection: ANSI Z89.1 Type 1 Class G; Face Shield/Eye Protection ANSI Z87.1+ :2015 Certified; SEI Certified and Listed |
|---|---|
| Assigned Protection Factor (APF) | 1000 |
| Battery Life | 7 hours with two batteries |
| Battery Type | 5 Cell Nickel Metal Hydride |
| Cartridge/Filter | PARTICULATE |
| Catalog Number | Mfr. No. | Model | Includes | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Model | Includes | Price | Quantity | ||||
18444068
|
Gentex™
PR05801B2A1 |
PR05801-B-2A1 | Black Stedprene FR Neck Cape, HE Filter, ADF Welding Lens, Internal Switch, Black Hard Hat |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
18444073
|
Gentex™
PR05801B2C1 |
PR05814-B-2A1 | Black Stedprene FR Neck Cape, HE Filter, ADF Welding Lens, Internal and External Switches, Black Hard Hat |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- No waist belts, hoses, or cables for maximum mobility
- High 8.1 CFM airflow rate
- Cinched Stedprene fire-resistant drawstring neck cape
- Hard Hat provides ANSI Z89.1 compliant head protection
- Choose from black or hi vis color options
- Quick release adjustable headband for optimal balance and fit
- Enhanced IP67 sealed blower/motor assembly
Specifications
| Head Protection: ANSI Z89.1 Type 1 Class G; Face Shield/Eye Protection ANSI Z87.1+ :2015 Certified; SEI Certified and Listed | |
| 7 hours with two batteries | |
| PARTICULATE | |
| PAPR System for Welding | |
| HEPA (HE) or HEPA/HF/HC *Nuisance OV | |
| Under 70 dB | |
| 2.5 hr. |
| 1000 | |
| 5 Cell Nickel Metal Hydride | |
| NIOSH Approved | |
| One Size | |
| 8.1 CFM or 230 L/min. | |
| 1 Ea. | |
| One Size |