missing translation for 'onlineSavingsMsg'
Learn More

Gentex™ PureFlo ESM+ PF60 Welding PAPR DFS Item

Head mounted self-contained PAPR ideal for welding, fabrication, and radiant heat hazards.

Supplier:  Gentex™ PR05801B2A1

 View more versions of this product

Catalog No. 18444068

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

  • No waist belts, hoses, or cables for maximum mobility
  • High 8.1 CFM airflow rate
  • Cinched Stedprene fire-resistant drawstring neck cape
  • Hard Hat provides ANSI Z89.1 compliant head protection
  • Choose from black or hi vis color options
  • Quick release adjustable headband for optimal balance and fit
  • Enhanced IP67 sealed blower/motor assembly
Specifications

Specifications

Head Protection: ANSI Z89.1 Type 1 Class G; Face Shield/Eye Protection ANSI Z87.1+ :2015 Certified; SEI Certified and Listed
7 hours with two batteries
PARTICULATE
PAPR System for Welding
HEPA (HE) or HEPA/HF/HC *Nuisance OV
Black Stedprene FR Neck Cape, HE Filter, ADF Welding Lens, Internal Switch, Black Hard Hat
Under 70 dB
2.5 hr.
1000
5 Cell Nickel Metal Hydride
NIOSH Approved
One Size
8.1 CFM or 230 L/min.
PR05801-B-2A1
1 Ea.
One Size
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions