missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gentex™ PureFlo ESM+ PF60 Welding PAPR
Head mounted self-contained PAPR ideal for welding, fabrication, and radiant heat hazards.
Supplier: Gentex™ PR05801B2C1
Description
- No waist belts, hoses, or cables for maximum mobility
- High 8.1 CFM airflow rate
- Cinched Stedprene fire-resistant drawstring neck cape
- Hard Hat provides ANSI Z89.1 compliant head protection
- Choose from black or hi vis color options
- Quick release adjustable headband for optimal balance and fit
- Enhanced IP67 sealed blower/motor assembly
Specifications
| Head Protection: ANSI Z89.1 Type 1 Class G; Face Shield/Eye Protection ANSI Z87.1+ :2015 Certified; SEI Certified and Listed | |
| 7 hours with two batteries | |
| PARTICULATE | |
| PAPR System for Welding | |
| HEPA (HE) or HEPA/HF/HC *Nuisance OV | |
| Black Stedprene FR Neck Cape, HE Filter, ADF Welding Lens, Internal and External Switches, Black Hard Hat | |
| Under 70 dB | |
| 2.5 hr. |
| 1000 | |
| 5 Cell Nickel Metal Hydride | |
| NIOSH Approved | |
| One Size | |
| 8.1 CFM or 230 L/min. | |
| PR05814-B-2A1 | |
| 1 Ea. | |
| One Size |