missing translation for 'onlineSavingsMsg'
Learn More

BIOTIX™ Pure xTIP Rainin LTS Compatible Racked Pipette Tips

Designed with Flexfit™ to lower necessary insertion and ejection force by ~70% to make secure seal. Manufactured with Pure resin made of virgin polypropylene.

$124.30 - $130.57

Specifications

Filtered Non-filtered
Color Clear
Autoclavable Autoclavable
Certifications/Compliance BSE/TSE Free, Endotoxins/Pyrogen Free, DNASE/RNASE Free, PCR Inhibitor Free
For Use With (Application) Manual Volume Transfer
View More Specs

Products 10
Catalog Number Mfr. No. Volume (Metric) Sterility Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Sterility Price Quantity  
12-578-070
View Documents Promotion Details
BIOTIX™
63305480
1200 μL Non-sterile
Pack of 768 Each for $124.30
Case of 5 Pack for $621.50
Only null left
 
12-578-071
View Documents Promotion Details
BIOTIX™
63305479
1200 μL Sterile
Pack of 768 Each for $130.57
Case of 5 Pack for $652.85
Only null left
 
12-578-073
View Documents Promotion Details
BIOTIX™
63305474
1000 μL Non-sterile
Pack of 768 Each for $124.30
Case of 5 Pack for $621.50
Only null left
 
12-578-074
View Documents Promotion Details
BIOTIX™
63305473
1000 μL Sterile
Pack of 768 Each for $130.57
Case of 5 Pack for $652.85
Only null left
 
12-578-075
View Documents Promotion Details
BIOTIX™
63305469
300 μL Non-sterile
Pack of 960 Each for $124.30
Case of 5 Pack for $621.50
Only null left
 
12-578-076
View Documents Promotion Details
BIOTIX™
63305468
300 μL Sterile
Pack of 960 Each for $130.57
Case of 5 Pack for $652.85
Only null left
 
12-578-078
View Documents Promotion Details
BIOTIX™
63305463
250 μL Non-sterile
Pack of 960 Each for $124.30
Case of 5 Pack for $621.50
Only null left
 
12-578-079
View Documents Promotion Details
BIOTIX™
63305462
250 μL Sterile
Pack of 960 Each for $130.57
Case of 5 Pack for $652.85
Only null left
 
12-578-081
View Documents Promotion Details
BIOTIX™
63305457
20 μL Non-sterile
Pack of 960 Each for $124.30
Case of 5 Pack for $621.50
Only null left
 
12-578-082
View Documents Promotion Details
BIOTIX™
63305456
20 μL Sterile
Pack of 960 Each for $130.57
Case of 5 Pack for $652.85
Only null left
 
Description

Description

  • Fits Rainin LTS and x Pipette pipettes
  • FlexFit technology features flexible proximal end to ensure secure seal with 66-80% less insertion force than competitor tips
  • Pure resin made of virgin polypropylene
  • Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip
  • Certified free of RNase, DNase, Pyrogens, and PCR Inhibitors
Specifications

Specifications

Non-filtered
Autoclavable
Manual Volume Transfer
Pipette Tips
Rainin LTS
xTIP 4
Standard
Clear
BSE/TSE Free, Endotoxins/Pyrogen Free, DNASE/RNASE Free, PCR Inhibitor Free
Ergonomic
Single-use
Yes
100% Virgin Polypropylene
Videos
SDS
Documents

Documents