Learn More
United Scientific™ Pulley
Ideal for use in demonstrating simple machines.
Supplier:  United Scientific™ PULT03
 View more versions of this product
Catalog No. S15840
Only null left
Description
Description
  • Free-running single pulley with 50 mm diameter plastic sheave.
  • Designed with deep V-groove, aluminum frame, and swivel hooks on both ends.
  • Available in single, double, triple, and quadruple pulleys.
Specifications
Chemical Identifiers
50 mm
Specifications
Triple Parallel Pulley
50 mm
Product Suggestions
Product Suggestions