missing translation for 'onlineSavingsMsg'
Learn More

Pterostilbene 98.0+%, TCI America™

Catalog No. P1924100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1924100MG 100 mg
P19241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1924100MG Supplier TCI America Supplier No. P1924100MG
Only null left

Chemical Identifiers

CAS 537-42-8
Molecular Formula C16H16O3
Molecular Weight (g/mol) 256.301
MDL Number MFCD00238710
InChI Key VLEUZFDZJKSGMX-ONEGZZNKSA-N
Synonym 4′C-Hydroxy-3,5-dimethoxy-trans-stilbene, trans-1-(3,5-Dimethoxyphenyl)-2-(4-hydroxyphenyl)ethylene
PubChem CID 5281727
ChEBI CHEBI:8630
IUPAC Name 4-[(E)-2-(3,5-dimethoxyphenyl)ethenyl]phenol
SMILES COC1=CC(=CC(=C1)C=CC2=CC=C(C=C2)O)OC

Specifications

Color White
Quantity 100 mg
Formula Weight 256.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pterostilbene
TSCA No
Recommended Storage Refrigerator