CAS RN 537-42-8
CAS RN 537-42-8
IUPAC Name:
4-[(1E)-2-(3,5-dimethoxyphenyl)ethenyl]phenol
Synonyms:
pterostilbene 3',5'-dimethoxy-4E-stilbenol trans-pterostilbene 3',5'-dimethoxy-resveratrol 3',5'-dimethoxy-4-stilbenol 3',5'-dimethoxy-4e-stilbenol resveratrol dimethyl ether
Molecular Weight (g/mol):
256.3
Molecular Formula:
C16H16O3
InChi Key:
VLEUZFDZJKSGMX-ONEGZZNKSA-N
SMILES:
COC1=CC(\C=C\C2=CC=C(O)C=C2)=CC(OC)=C1
1
–
1
of
1
results
1
Pterostilbene 98.0+%, TCI America™
CAS: 537-42-8 Molecular Formula: C16H16O3 Molecular Weight (g/mol): 256.301 MDL Number: MFCD00238710 InChI Key: VLEUZFDZJKSGMX-ONEGZZNKSA-N Synonym: 4′C-Hydroxy-3,5-dimethoxy-trans-stilbene, trans-1-(3,5-Dimethoxyphenyl)-2-(4-hydroxyphenyl)ethylene PubChem CID: 5281727 ChEBI: CHEBI:8630 IUPAC Name: 4-[(E)-2-(3,5-dimethoxyphenyl)ethenyl]phenol SMILES: COC1=CC(=CC(=C1)C=CC2=CC=C(C=C2)O)OC