missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ PSC 833

Catalog No. 40421
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40421 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40421 Supplier R&D Systems™ Supplier No. 4042/1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Inhibitor of P-gp-mediated MDR

P-glycoprotein (P-gp) modulator; inhibits P-gp-mediated multidrug-resistance (MDR). Reverses resistance to several cytotoxic drugs including mitoxantrone and doxorubicin (resistance factors are 2.0 and 6.5 respectively) in human MDR cancer cell lines. Non-immunosuppressive analog of cyclosporin A (Cat. No. 1101).

Chemical Identifiers

CAS 121584-18-7
Molecular Formula C63H111N11O12

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 1214.62
Purity >98%
Molecular Weight (g/mol) 1214.65
Chemical Name or Material PSC 833
Recommended Storage Store at -20°C