missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PSB 603

Catalog No. 319810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
319810 10 mg
319850 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 319810 Supplier Tocris Bioscience™ Supplier No. 3198/10

Highly selective A2B antagonist

Adenosine A2B receptor antagonist that displays > 17000-fold selectivity over other adenosine receptors (Ki values are 0.553, > 10000, > 10000 and > 10000 nM for A2B, A1, A2A and A3 receptors respectively).

Chemical Identifiers

CAS 1092351-10-4
Molecular Formula C24H23ClN6O4S

Specifications

Quantity 10 mg
Formula Weight 529.01
Purity >98%
Molecular Weight (g/mol) 526.996
Chemical Name or Material PSB 603
Recommended Storage Store at +4°C