missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PSB 0788

Catalog No. 319910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
319910 10 mg
319950 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 319910 Supplier Tocris Bioscience™ Supplier No. 3199/10

Selective A2B antagonist

Potent and selective adenosine A2B receptor antagonist (IC50 = 3.64 nM at human A2B receptors). Displays > 100-fold selectivity over hA1, hA2A and hA3 receptors.

Chemical Identifiers

CAS 1027513-54-7
Molecular Formula C25H27ClN6O4S

Specifications

Quantity 10 mg
Formula Weight 543.04
Purity >98%
Molecular Weight (g/mol) 543.039
Chemical Name or Material PSB 0788
Recommended Storage Store at +4°C