missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ PRP™-X300 Ion Exclusion HPLC Columns

Catalog No. 1481638
Change view
Click to view available options
Length (Metric):
250 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
1481638 250 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1481638 Supplier Hamilton™ Supplier No. HAM79465

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Polymeric column packing for the separation of alcohols and organic acids

A fast, versatile HPLC column packing material, Hamilton PRP-X300 columns rapidly and easily separate closely related alcohols and organic acids without column heating. The sulfonated poly(styrene-divinylbenzene) support separates samples by a mixed mode mechanism.
Three Separation Modes

  • Hydrogen Bonding, Reversed Phase, and Ion Exclusion
  • Wide Applicability (Versatility)
    • A wide range of samples can be analyzed with PRP-X300 columns because the selectivity of the column can be altered by changing the pH of the buffer or adding an organic modifier (Methanol, Acetonitrile) to the buffer. The supports stability to organic solvents makes it possible to analyze samples that have been too highly retained on conventional ion exclusion supports. The use of isocratic conditions allows samples to be analyzed without column reequilibration.
  • Fast Results
    • The excellent selectivity of the PRP-X300 columns ensures most separations are complete within five minutes
  • Easy Sample Preparation
    • Most samples require only minimal preparation before injection

    • One particle size: 7μm
    • Three column internal diameters: 1.0 to 4.6mm
    • Two column materials: 316 stainless steel and PEEK
    • Analytical and semiprep/preparative guard columns

    Specifications

    Column Format Analytical Column
    Diameter (Metric) 4.1 mm
    Length (Metric) 250 mm
    Max. Pressure 5000 psi (345 bar)
    Mobile Phase Compatibility pH 1 to 13, 0 to 100% aqueous, organic modifier
    Particle Size 7 μm
    pH 1 to 13
    Pore Size 100 Å
    Temperature 5°C to 60°C
    USP Type L17
    Stationary Phase PRP-X300
    Product Line PRP-X300
    Packing Material PSDVB/Sulfonic Acid
    Column Type Ion Exchange
    Phase Ion Exchange
    Material Stainless Steel
    Technique Mixed Mode
    Product Type Column
    Show More Show Less