Learn More
MilliporeSigma™ Supelco™ Proteomix™ SCX-NP5 Strong Cation Exchange Column, 5 μm Particle Size, Stainless Steel
Used for the analysis or small scale purification of positively charged biopolymers including glycoprotein isoforms and mAb variants.
$1844.46 - $2795.62
Specifications
Particle Size 5 μm
pH 2.0 to 12.0
Pore Size Nonporous
Stationary Phase Sulfonic Acid
Product Line Proteomix
View More Specs
Products 4
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Price Quantity  
111006912
View Documents
MilliporeSigma™ Supelco™
Z777156
5 cm 2.1 mm
Each for $1,844.46
Only null left
111006398
View Documents
MilliporeSigma™ Supelco™
Z777157
15 cm 2.1 mm
Each for $2,795.62
Only null left
111006541
View Documents
MilliporeSigma™ Supelco™
Z777159
5 cm 4.6 mm
Each for $1,844.46
Only null left
111006399
View Documents
MilliporeSigma™ Supelco™
Z777160
15 cm 4.6 mm
Each for $2,795.62
Only null left
Description
Description
Proteomix™ columns are packed with poly(styrene divinylbenzene) particles that are shielded by a hydrophilic polymer layer that contains a network of weak (WAX) or strong (SAX) anion exchange groups.
Specifications
Specifications
5 μm
Nonporous
Proteomix
Cation Exchange
Cation Exchange
2.0 to 12.0
Sulfonic Acid
Spherical PS-DVB/Sulfonic Acid Phase
Stainless Steel
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Proteomix is a registered trademark of Sepax Technologies