missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Proteomix™ HIC Propyl-NP5 Column, 5 μm Particle Size

Catalog No. 111006521
Change view
Click to view available options
Length (Metric):
35 mm
50 mm
100 mm
Diameter (Metric):
4.6 mm
7.8 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
111006521 35 mm 4.6 mm
111006391 100 mm 4.6 mm
111006392 50 mm 7.8 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111006521 Supplier MilliporeSigma™ Supelco™ Supplier No. 61881U

Proteomix™ HIC columns are specially designed for high resolution and high efficiency separations of proteins, oligonucleotides and peptides.

Utilizing proprietary surface technologies, Proteomix HIC-NP resin is made of non-porous polystyrenedivinylbenzene (PS/DVB) beads with narrow-dispersed particle size distribution. The PS/DVB bead is modified with alkyl groups or aryl group that provides hydrophobic interaction with analytes. Proteomix HIC-NP resin is highly rigid and mechanically stable. In comparison to silica based HIC phase media, Proteomix HIC-NP phases have advantages for biomolecule separations with wide pH range (2-12) and high thermal stability. The nonporous structure and narrow particle distribution offer special selectivity, high resolution separation of proteins such as mAb (monoclonal antibody), ADC (antibody drug conjugate) and related protein fragments, DNA and oligonucleotides. Proteomix HIC-NP media is applicable at laboratory discovery, laboratory-scale purification and process chromatography for the production of a few mgs to kilogram of proteins.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 35 mm
Particle Size 5 μm
pH 2 - 12
Pore Size Nonporous
Stationary Phase Propyl
Product Line Proteomix
Matrix Propyl Phase/Polystyrene/Divinylbenzene Particle Platform (Spherical, highly-crossed linked)
Phase Hydrophobic Interaction
Material Stainless Steel Hardware
Technique HIC HPLC
Show More Show Less

Proteomix is a registered trademark of Sepax Technologies