missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Proteinase K Solution, 600mAU/mL

Catalog No. 710494
Change view
Click to view available options
Quantity:
2 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
710494 10 mL
710493 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 710494 Supplier MilliporeSigma™ Supplier No. 710494

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Efficient removal of proteins from nucleic acid solutions

  • Proteinase K is a highly active 28,904-Da serine protease isolated from the fungus Tritirachium album 
  • The enzyme exhibits broad cleavage specificity on native and denatured proteins and is widely used in the purification of DNA and RNA
  • Its activity is increased in the presence of denaturants such as SDS (1%) and elevated temperature (50-60°C)
  • The recommended working concentration is 50-100μg/mL for protein removal and enzyme inactivation, and up to 2mg/mL for tissue treatment
  • The Proteinase K, Lyophilized powder can be prepared as a 20mg/mL stock solution in water and stored in aliquots at -20°C
  • The enzyme is also available as a ready-to-use concentrated stock solution (600mAU/mL) that is convenient for routine use in most applications
  • 1 mg of Proteinase K is the equivalent of 30mAU (AU = Anson unit)
  • The Novagen Proteinase K products are free of detectable DNase and RNase

Specifications

Content And Storage -20°C
Quantity 10 mL
Product Type Proteinase K