Learn More
Thermo Scientific™ Proteinase K, recombinant, PCR grade
Degrade proteins even in the presence of detergents with Proteinase K, a broad-range endolytic protease widely used for digestion of proteins in nucleic acid preparations.
$112.13 - $413.22
Specifications
Product Type Proteinase K
Form Liquid
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FEREO0491
View Documents Promotion Details
Thermo Scientific™
EO0491
1 mL
Each for $112.13
Only null left
FEREO0492
View Documents Promotion Details
Thermo Scientific™
EO0492
5 x 1.0 mL
Each for $413.22
Only null left
Description
Description
Proteinase K is an endolytic protease that cleaves peptide bonds at the carboxylic sides of aliphatic, aromatic, or hydrophobic amino acids. The Proteinase K is classified as a serine protease. The smallest peptide to be hydrolyzed by this enzyme is a tetrapeptide.
Highlights
  • Ready-to-use solution
  • Active in a wide range of reaction conditions
Specifications
Specifications
Proteinase K
Liquid
Videos
SDS
Documents
Documents
For Research Use Only. Not for use in diagnostic procedures.