missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Proteinase K, Lyophilized

Catalog No. p-6814334
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
706635 500 mg
706634 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 706635 Supplier MilliporeSigma™ Supplier No. 706635
Only null left

Highly active serine protease that exhibits broad cleavage specificity on native and denatured proteins and is widely used in the purification of DNA and RNA

  • Proteinase K is a highly active 28,904-Da serine protease isolated from the fungus Tritirachium album
  • Its activity is increased in the presence of denaturants such as SDS (1%) and elevated temperature (50-60°C)
  • The recommended working concentration is 50-100μg/mL for protein removal and enzyme inactivation, and up to 2mg/mL for tissue treatment
  • The Proteinase K, Lyophilized powder can be prepared as a 20mg/mL stock solution in water and stored in aliquots at -20°C
  • The enzyme is also available as a ready-to-use concentrated stock solution (600mAU/mL) that is convenient for routine use in most applications
  • 1mg of Proteinase K is the equivalent of 30mAU (AU = Anson unit)
  • The Novagen Proteinase K products are free of detectable DNase and RNase

Specifications

CAS 39450-01-6
Content And Storage 2°C to 8°C
Quantity 500 mg
Product Type Proteinase K