Learn More
Invitrogen™ eBioscience™ Protein Transport Inhibitor Cocktail (500X)
Catalog No. p-7091854
Change view
Click to view available options
Quantity:
100 μL
4 x 100 μL
Catalog No. Quantity
509309 100 μL
501129032 4 x 100 μL
2 options
Catalog No. 509309
Supplier: Invitrogen™ 00498093
Only null left
Suitable for use in assays in which prevention of protein transport to the extracellular space is desired.
  • A cocktail of Brefeldin A and Monensin
  • Suitable for use in assays in which prevention of protein transport to the extracellular space is desired
  • It has been specifically formulated for use as a negative control for the Cell Stimulation Cocktail (plus protein transport inhibitors) (500X)
  • Addition of this cocktail to cells in culture inhibits the intracellular protein secretory/transport pathway resulting in the accumulation of secreted proteins in the lumen of the endoplasmic reticulum and in the Golgi apparatus
  • This accumulation can then be detected by intracellular staining and flow cytometric analysis or western blotting
Specifications
Product Line eBioscience
Type Protein Transport Inhibitor Cocktail
Quantity 100 μL
For Research Use Only. Not for use in diagnostic procedures.