Learn More
Invitrogen™ eBioscience™ Protein Transport Inhibitor Cocktail (500X)
Catalog No. 501129032
Change view
Click to view available options
Quantity:
100 μL
4 x 100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501129032 4 x 100 μL
509309 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 501129032 Supplier Invitrogen™ Supplier No. 00498003
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Suitable for use in assays in which prevention of protein transport to the extracellular space is desired.
  • A cocktail of Brefeldin A and Monensin
  • Suitable for use in assays in which prevention of protein transport to the extracellular space is desired
  • It has been specifically formulated for use as a negative control for the Cell Stimulation Cocktail (plus protein transport inhibitors) (500X)
  • Addition of this cocktail to cells in culture inhibits the intracellular protein secretory/transport pathway resulting in the accumulation of secreted proteins in the lumen of the endoplasmic reticulum and in the Golgi apparatus
  • This accumulation can then be detected by intracellular staining and flow cytometric analysis or western blotting
Specifications
Product Line eBioscience
Type Protein Transport Inhibitor Cocktail
Quantity 4 x 100 μL
For Research Use Only. Not for use in diagnostic procedures.