missing translation for 'onlineSavingsMsg'
Learn More

Cytiva™ HiTrap™ Protein Select™ Column

Catalog No. 09928218
Change view
Click to view available options
Volume (Metric):
1 x 1 mL
1 x 5 mL
5 x 1 mL
5 x 5 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
09928218 5 x 5 mL
09928217 1 x 5 mL
09928220 5 x 1 mL
09928219 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 09928218 Supplier Cytiva™ Supplier No. 17542154
Only null left

An affinity chromatography resin for purifying recombinant proteins using the self-cleaving Cytiva™ Protein Select™ tag.

Cytiva™ Protein Select™ resin is an affinity chromatography resin for purifying any recombinant protein that does not have an affinity binding partner.

  • Simple affinity purification protocol for recombinant proteins.
  • One-step purification and tag cleavage without the need for proteases or other elution reagents.
  • Delivers a highly pure, native protein with no remaining tag amino acids after tag cleavage.
  • Can be processed under mild conditions, which means no exposure to conditions that cause protein aggregation or loss of biological activity.
  • Flexible, enabling the use in multiple expression systems.

Specifications

Type Column
Quantity 5 x 5 mL
Form Liquid
Column Type Pre-packed Column
Flow Rate 20 mL/min.
Volume (Metric) 5 x 5 mL
Diameter (Metric) 16 mm
Length (Metric) 25 mm