missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva™ HiTrap™ Protein Select™ Column
An affinity chromatography resin for purifying recombinant proteins using the self-cleaving Cytiva™ Protein Select™ tag.
$309.01 - $6359.53
Specifications
| Type | Column |
|---|---|
| Form | Liquid |
| Column Type | Pre-packed Column |
| Length (Metric) | 25 mm |
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity | |||||
|
09-928-217
|
Cytiva™
17542153 |
1 x 5 mL |
Each for $1,545.05
|
|
|||||
|
09-928-218
|
Cytiva™
17542154 |
5 x 5 mL |
Each for $6,359.53
|
|
|||||
|
09-928-220
|
Cytiva™
17542152 |
5 x 1 mL |
Each for $1,545.05
|
|
|||||
|
09-928-219
|
Cytiva™
17542151 |
1 x 1 mL |
Each for $309.01
|
|
|||||
Description
Cytiva™ Protein Select™ resin is an affinity chromatography resin for purifying any recombinant protein that does not have an affinity binding partner.
- Simple affinity purification protocol for recombinant proteins.
- One-step purification and tag cleavage without the need for proteases or other elution reagents.
- Delivers a highly pure, native protein with no remaining tag amino acids after tag cleavage.
- Can be processed under mild conditions, which means no exposure to conditions that cause protein aggregation or loss of biological activity.
- Flexible, enabling the use in multiple expression systems.
Specifications
| Column | |
| Liquid | |
| Pre-packed Column | |
| 25 mm |