Learn More
Promega Protein Precipitation Solution for Wizard™ Genomic DNA Purification Kits
An easy, solution-based method for isolation of DNA from white blood cells, tissue culture cells, animal tissue, plant tissue, yeast and Gram-positive and Gram-negative bacteria.
Supplier: Promega A7953
Description
- Manual, Solution-Based DNA Extraction Method
- DNA from blood, animal and plant cells, bacteria or yeast
- 60-minute, scalable protocol
- Kit size based on number of isolations from whole blood, other sample types may vary
Specifications
| 15°C to 30°C | |
| Isolation of DNA from white Blood cells, tissue culture cells, animal tissue, plant tissue, yeast and gram positive and gram negative bacteria | |
| 350 mL |
| Wizard™ genomic DNA purification kit | |
| Protein Precipitation Solution |