missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Protein Kinase Inhibitor
$543.18
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ61687MCR
|
Thermo Scientific Chemicals
J61687MCR |
1 mg |
Each for $543.18
|
|
|||||
Description
Inhibitor of protein kinase A
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 97161-97-2 | |
| C27H21N3O5 | |
| K252a |
Specifications
| Off-white | |
| Inhibitor of protein kinase A | |
| C27H21N3O5 | |
| K252a | |
| KOZFSFOOLUUIGY-UHFFFAOYNA-N | |
| methyl 16-hydroxy-15-methyl-3-oxo-28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8(13),9,11,20,22,24-nonaene-16-carboxylate |
| -30°C to -10°C | |
| 97161-97-2 | |
| 467.47 | |
| Powder | |
| COC(=O)C1(O)CC2OC1(C)N1C3=C(C=CC=C3)C3=C1C1=C(C4=CC=CC=C4N21)C1=C3CNC1=O | |
| 467.48 |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only