CAS RN 97161-97-2
IUPAC Name:
methyl 16-hydroxy-15-methyl-3-oxo-28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8(13),9,11,20,22,24-nonaene-16-carboxylate
Synonyms:
staurosporinone
Molecular Weight (g/mol):
467.48
Molecular Formula:
C27H21N3O5
InChi Key:
KOZFSFOOLUUIGY-UHFFFAOYNA-N
SMILES:
COC(=O)C1(O)CC2OC1(C)N1C3=C(C=CC=C3)C3=C1C1=C(C4=CC=CC=C4N21)C1=C3CNC1=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals Protein Kinase Inhibitor
CAS: 97161-97-2 Molecular Formula: C27H21N3O5,C27H21N3O5 Synonym: K252a