missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Protein G Sepharose™ 4 Fast Flow Media

Catalog No. 45000116
Change view
Click to view available options
Quantity:
5 mL
25 mL
200 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45000116 5 mL
45000117 25 mL
45000118 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45000116 Supplier Cytiva Supplier No. 17061801

Rapid processing of large volumes, recoveries of 70 to 90%. Cytiva Protein G Sepharose™ 4 Fast Flow Media is recombinant protein G coupled to Sepharose 4 Fast Flow.

  • Binding specificities that complement Protein A media
  • Binds a broad range of IgG species and subclasses
  • High capacity, >20mg human IgG/mL medium; high flowrate
  • Multi-point attachment minimizes ligand leakage
  • Used in a range of diagnostic and research applications

Specifications

Ligand Concentration 2mg Protein G/mL Medium
Matrix 4% Cross Linked Agarose
Particle Size 90 μm
pH 3 to 9
Quantity 5 mL
Content And Storage 2°C to 8°C
For Use With (Application) Used in a range of diagnostic and research applications
Ligand Recombinant Protein G lacking albumin binding region