Learn More
Applied Biosystems™ Protein Expression Sample Prep Kit
Catalog No. 501148551
Change view
Click to view available options
Quantity:
1000 reactions
Catalog No. Quantity
501148551 1000 reactions
1 options
Catalog No. 501148551 Supplier Applied Biosystems™ Supplier No. 4405443
Season of Savings
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Protein Expression Sample Prep Kit
The Applied Biosystems™ Protein Expression Sample Preparation Kit contains the reagents required for the preparation of target and control cell lysates for use with TaqMan™ Protein Expression Assay experiments. The Cell Lysis Reagent is a buffered detergent solution, which is combined with user-supplied protease and phosphatase inhibitors, and used to lyse adherent or nonadherent cultured cells under nondenaturing conditions. The Cell Resuspension Buffer is used to resuspend the cells before adding the lysis reagent and to dilute the lysates in preparation for the assay-binding step.
Specifications
Type Cell Lysis Buffer
Quantity 1000 reactions
Content And Storage Contains Cell Lysis Reagent (2x) (3 x 1 mL tubes) and Cell Resuspension Buffer (1 x 25 mL bottle). Store at 2 to 8°C.
Product Line TaqMan
For Research Use Only. Not for use in diagnostic procedures.