missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Protein A and Protein G Mag Sepharose™

Catalog No. 45002161
Change view
Click to view available options
Quantity:
500 μL
4 x 500 μL
Binding Capacity:
13 to 22mg Human IgG/mL
8 to 17mg Human IgG/mL
Ligand:
Native Protein A
Protein G
Bead Type:
Protein A Mag Sepharose
Protein G Mag Sepharose
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Binding Capacity Ligand Bead Type
45002161 500 μL 8 to 17mg Human IgG/mL Native Protein A Protein A Mag Sepharose
45002160 500 μL 13 to 22mg Human IgG/mL Protein G Protein G Mag Sepharose
45002173 4 x 500 μL 13 to 22mg Human IgG/mL Protein G Protein G Mag Sepharose
45002174 4 x 500 μL 8 to 17mg Human IgG/mL Native Protein A Protein A Mag Sepharose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 45002161 Supplier Cytiva Supplier No. 28944006

Simplify enrichment of target proteins by immunoprecipitation or pull-down applications

Protein A and Protein G Mag Sepharose are magnetic beads designed to simplify enrichment of target proteins by immunoprecipitation or pull-down applications. The magnetic beads are based on Sepharose with native protein A or native protein G as ligand. Protein A and Protein G have high affinities for the Fc region of IgG antibodies and offer differing affinity characteristics across a broad range of mammalian IgG species.

There are two protocols available for protein enrichment: the cross-link protocol and the classic protocol. The protocols are simple with elution conditions optimized for for downstream applications such as electrophoresis and MS.

Protein A and Protein G Mag Sepharose combine well-proven enrichment methods with the magnetic beads platform, which has ideal properties for small-scale sample preparation. Together with MagRack 6, a separation tool for six microcentrifuge tubes, up to six samples can be processed in parallel.

  • Visible and dense Sepharose based magnetic beads with protein A or protein G ligand, easy to spot and collect bound target protein
  • Non-adherent beads eliminate smearing effects and aggregate formation. Can be used without detergents
  • Simple capture of target protein in small or large sample volumes (low microliter to high milliliter scale)
  • Optimized capacity for enrichment/immunoprecipitation of target proteins - only low amounts of antibody needed

Matrix: Paramagnetic, spherical, highly cross-linked agarose particles
Ligand: Native protein A; Native protein G
Binding capacity Protein A: 8-17mg human IgG/mL gel
Binding capacity Protein G: 13-22mg human IgG/mL gel
Particle size: 37-100μm
Working temperature: Room temperature
Storage solution: 20% ethanol
Storage temperature: 4°C to 8°C

Specifications

Bead Type Protein A Mag Sepharose
Binding Capacity 8 to 17mg Human IgG/mL
Matrix Paramagnetic spherical, highly cross linked agarose particles
Particle Size 37 to 100 μm
Quantity 500 μL
Content And Storage 4°C to 8°C
For Use With (Application) Designed to simplify enrichment of target proteins by immunoprecipitation or pull-down applications, for biomarker research
Ligand Native Protein A