missing translation for 'onlineSavingsMsg'
Learn More

Promega ProteaseMAX™ Surfactant, Trypsin Enhancer DFS Item

ProteaseMAX Surfactant, Trypsin Enhancer, enhances the enzymatic performance of Trypsin, Chymotrypsin and Lys-C in digestion reactions.

$124.76 - $538.89

Specifications

Formulation Lyophilized
Product Type Trypsin Enhancer
Content And Storage -30°C to -10°C
For Use With (Application) Protein isolation and purification
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRV2072 DFS Item
View Documents
Promega
V2072
5 x 1 mg
Each for $538.89
Only null left
 
PRV2071 DFS Item
View Documents
Promega
V2071
1 mg
Each for $124.76
Only null left
 
Description

Description

  • Improved In-Gel and In-Solution Protein Digestion
  • Enhances enzymatic performance of trypsin, chymotrypsin and Lys-C
  • Saves time and labor when used with in-gel protein digestion
  • Complete digestion in 1 hour
Specifications

Specifications

Lyophilized
Trypsin Enhancer
-30°C to -10°C
Protein isolation and purification
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'