missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Protease QF

Catalog No. QVT0512
Change view
Click to view available options
Quantity:
180 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QVT0512 180 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. QVT0512 Supplier Invitrogen™ Supplier No. QVT0512

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protease QF, 180 μL, is an enzyme in aqueous buffered solution and an individual component of the ViewRNA™ ISH Tissue Assay Kits, 2-Plex, 24 assays.

Protease QF, 180 μL, is an enzyme in aqueous buffered solution and an individual component of the ViewRNA™ ISH Tissue Assay Kits, 2-Plex, 24 assays.

Specifications

Content And Storage Store at 2°C to 8°C

IMPORTANT! Do not freeze.
Form Liquid
Quantity 180 μL
Product Type Protease

For Research Use Only. Not for use in diagnostic procedures.