missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protease from Aspergillus oryzae, TCI America™
Specifications
Specifications
| Color | Pale yellow - Grayish yellow red at 20°C |
| Content And Storage | 0°C to 10°C |
| Quantity | 25 g |
| Form | Crystalline Powder |