missing translation for 'onlineSavingsMsg'
Learn More

Propylparaben, NF, BP, EP, 98-102%, Spectrum™ Chemical

Catalog No. 18605935
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605935 500 g Poly Bottle
18605936 2.5 kg Poly Bottle
18605937 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18605935 Supplier Spectrum Chemical Mfg Cor Supplier No. PR133500GM

PR133, 94-13-3, C10H12O3

Spectrum™ Chemical Propylparaben, NF, BP, EP is used as an antimicrobial preservative. All Spectrum Chemical NF, BP, EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 94-13-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
InChI Key QELSKZZBTMNZEB-UHFFFAOYSA-N
IUPAC Name propyl 4-hydroxybenzoate
SMILES CCCOC(=O)C1=CC=C(O)C=C1

Specifications

Acidity 0.001
CAS Max % 1
Ignition Residue 0.001
Melting Range 96-99°C
Packaging Poly Bottle
Quantity 500 g
Percent Purity 98 to 102%
Grade Multi Compendia
Physical Form Crystalline Powder or Lumps