missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propylparaben, FCC, 99-100.5%, Spectrum™ Chemical
P1452, 94-13-3, C10H12O3
Supplier: Spectrum Chemical Mfg Cor P145212KG
Description
Spectrum™ Chemical Propylparaben, FCC is used as an antimicrobial preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 94-13-3 | |
| 0.0005 | |
| 95-98°C | |
| C10H12O3 | |
| QELSKZZBTMNZEB-UHFFFAOYSA-N | |
| propyl 4-hydroxybenzoate | |
| 99 to 100.5% | |
| Crystalline Powder or Lumps |
| 1 | |
| 0.005 | |
| Poly Bottle | |
| 12 kg | |
| CCCOC(=O)C1=CC=C(O)C=C1 | |
| 180.20 | |
| FCC |
Chemical Identifiers
| 94-13-3 | |
| 180.20 | |
| propyl 4-hydroxybenzoate |
| C10H12O3 | |
| QELSKZZBTMNZEB-UHFFFAOYSA-N | |
| CCCOC(=O)C1=CC=C(O)C=C1 |