missing translation for 'onlineSavingsMsg'
Learn More

Propylparaben, FCC, 99-100.5%, Spectrum™ Chemical

Catalog No. 18605930
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605930 500 g Poly Bottle
18605931 2.5 kg Poly Bottle
18605932 12 kg Poly Bottle
18605933 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605930 Supplier Spectrum Chemical Mfg Cor Supplier No. P1452500GM

P1452, 94-13-3, C10H12O3

Spectrum™ Chemical Propylparaben, FCC is used as an antimicrobial preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 94-13-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
InChI Key QELSKZZBTMNZEB-UHFFFAOYSA-N
IUPAC Name propyl 4-hydroxybenzoate
SMILES CCCOC(=O)C1=CC=C(O)C=C1

Specifications

CAS Max % 1
Ignition Residue 0.0005
Loss on Drying 0.005
Melting Range 95-98°C
Packaging Poly Bottle
Quantity 500 g
Percent Purity 99 to 100.5%
Grade FCC
Physical Form Crystalline Powder or Lumps